missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl acetate, 99%

Catalog No. p-7026923
Change view
Click to view available options
Quantity:
250 mL
1000 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB23257AK 250 mL
AAB23257AU 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB23257AK Supplier Thermo Scientific Chemicals Supplier No. B23257AK
Only null left

CAS: 103-09-3 | C10H20O2 | 172.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 103-09-3
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.27
MDL Number MFCD00027249
InChI Key WOYWLLHHWAMFCB-UHFFFAOYNA-N
Synonym acetic acid, 2-ethylhexyl ester, 2-ethyl-1-hexanol acetate, 2-ethylhexyl ethanoate, 2-ethyl-1-hexyl acetate, 2-ethylhexanyl acetate, 2-ethylhexylacetate, beta-ethylhexyl acetate, acetic acid 2-ethylhexyl ester, fema number 2806, 2-ethylhexylester kyseliny octove
PubChem CID 7635
ChEBI CHEBI:87392
IUPAC Name 2-ethylhexyl acetate
SMILES CCCCC(CC)COC(C)=O

Specifications

Melting Point -45°C
Density 0.87
Boiling Point 199°C to 200°C
Flash Point 76°C (169°F)
Odor Pleasant
Refractive Index 1.42
Quantity 250 mL
Beilstein 1758321
Merck Index 14,6763
Formula Weight 172.27
Percent Purity 99%
Chemical Name or Material 2-Ethylhexyl acetate
Show More Show Less
Hazard Category H227-H315-H319
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
EINECSNumber 203-079-1
RTECSNumber AH5600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only