missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl 4-Methoxycinnamate 97.0+%, TCI America™

Supplier:  TCI America M1082100ML

 View more versions of this product

Catalog No. M1082100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

2-Ethylhexyl 4-Methoxycinnamate
Yellow
MFCD00072582
bidd:er0152, octyl methoxy cinnamate omc, unii-4y5p7mud51 component, 2s-2-ethylhexyl 2e-3-4-methoxyphenyl prop-2-enoate
CCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1
290.40
290.40
FCC
5466-77-3
C18H26O3
100 mL
YBGZDTIWKVFICR-UHFFFAOYNA-N
2-ethylhexyl 3-(4-methoxyphenyl)prop-2-enoate
11044481
≥97.0% (GC)
Liquid

Chemical Identifiers

5466-77-3
290.40
YBGZDTIWKVFICR-UHFFFAOYNA-N
11044481
CCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1
C18H26O3
MFCD00072582
bidd:er0152, octyl methoxy cinnamate omc, unii-4y5p7mud51 component, 2s-2-ethylhexyl 2e-3-4-methoxyphenyl prop-2-enoate
2-ethylhexyl 3-(4-methoxyphenyl)prop-2-enoate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1706

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions