missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl 4-Methoxycinnamate 97.0+%, TCI America™

Catalog No. p-7132528
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M108225ML 25 mL
M1082100ML 100 mL
M1082500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M108225ML Supplier TCI America Supplier No. M108225ML
Only null left

Chemical Identifiers

CAS 5466-77-3
Molecular Formula C18H26O3
Molecular Weight (g/mol) 290.40
MDL Number MFCD00072582
InChI Key YBGZDTIWKVFICR-UHFFFAOYNA-N
Synonym bidd:er0152, octyl methoxy cinnamate omc, unii-4y5p7mud51 component, 2s-2-ethylhexyl 2e-3-4-methoxyphenyl prop-2-enoate
PubChem CID 11044481
IUPAC Name 2-ethylhexyl 3-(4-methoxyphenyl)prop-2-enoate
SMILES CCCCC(CC)COC(=O)C=CC1=CC=C(OC)C=C1

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 290.40
Percent Purity ≥97.0% (GC)
Grade FCC
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl 4-Methoxycinnamate
EINECSNumber (3)-1706
TSCA Yes