missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylhexyl 3-Mercaptopropionate 98.0+%, TCI America™
$68.97 - $244.19
Chemical Identifiers
| CAS | 50448-95-8 |
|---|---|
| Molecular Formula | C11H22O2S |
| Molecular Weight (g/mol) | 218.355 |
| MDL Number | MFCD00087933 |
| InChI Key | SUODCTNNAKSRHB-UHFFFAOYSA-N |
| Synonym | 3-Mercaptopropionic Acid 2-Ethylhexyl Ester, 3-Mercaptopropionic Acid Octyl Ester, 2-Octyl 3-Mercaptopropionate |
| PubChem CID | 92640 |
| IUPAC Name | 2-ethylhexyl 3-sulfanylpropanoate |
| SMILES | CCCCC(CC)COC(=O)CCS |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
M106725ML
|
TCI America
M106725ML |
25 mL |
Each for $68.97
|
|
|||||
|
M1067500ML
|
TCI America
M1067500ML |
500 mL |
Each for $244.19
|
|
|||||
Chemical Identifiers
| 50448-95-8 | |
| 218.355 | |
| SUODCTNNAKSRHB-UHFFFAOYSA-N | |
| 92640 | |
| CCCCC(CC)COC(=O)CCS |
| C11H22O2S | |
| MFCD00087933 | |
| 3-Mercaptopropionic Acid 2-Ethylhexyl Ester, 3-Mercaptopropionic Acid Octyl Ester, 2-Octyl 3-Mercaptopropionate | |
| 2-ethylhexyl 3-sulfanylpropanoate |
Specifications
| 50448-95-8 | |
| C11H22O2S | |
| 25 mL | |
| SUODCTNNAKSRHB-UHFFFAOYSA-N | |
| 2-ethylhexyl 3-sulfanylpropanoate | |
| 92640 | |
| ≥98.0% (GC) | |
| 2-Ethylhexyl 3-Mercaptopropionate |
| Yellow | |
| MFCD00087933 | |
| 3-Mercaptopropionic Acid 2-Ethylhexyl Ester, 3-Mercaptopropionic Acid Octyl Ester, 2-Octyl 3-Mercaptopropionate | |
| CCCCC(CC)COC(=O)CCS | |
| 218.355 | |
| 218.36 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-1383
TSCA : Yes