missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethylhexyl 3-Mercaptopropionate 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Quantity | 500 mL |
| Formula Weight | 218.36 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Ethylhexyl 3-Mercaptopropionate |
Safety and Handling
| EINECSNumber | (2)-1383 |
| TSCA | Yes |