missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl 3-Mercaptopropionate 98.0+%, TCI America™

Catalog No. M106725ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M106725ML 25 mL
M1067500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M106725ML Supplier TCI America Supplier No. M106725ML
Only null left

Chemical Identifiers

CAS 50448-95-8
Molecular Formula C11H22O2S
Molecular Weight (g/mol) 218.355
MDL Number MFCD00087933
InChI Key SUODCTNNAKSRHB-UHFFFAOYSA-N
Synonym 3-Mercaptopropionic Acid 2-Ethylhexyl Ester, 3-Mercaptopropionic Acid Octyl Ester, 2-Octyl 3-Mercaptopropionate
PubChem CID 92640
IUPAC Name 2-ethylhexyl 3-sulfanylpropanoate
SMILES CCCCC(CC)COC(=O)CCS

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 218.36
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl 3-Mercaptopropionate
EINECSNumber (2)-1383
TSCA Yes