missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexyl 2-Cyano-3,3-diphenylacrylate 98.0+%, TCI America™

Catalog No. C096825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C096825G 25 g
C0968100G 100 g
C0968500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C096825G Supplier TCI America Supplier No. C096825G

Chemical Identifiers

CAS 6197-30-4
Molecular Formula C24H27NO2
Molecular Weight (g/mol) 361.49
MDL Number MFCD00059260
InChI Key FMJSMJQBSVNSBF-UHFFFAOYNA-N
Synonym 2-Cyano-3,3-diphenylacrylic Acid 2-Ethylhexyl Ester, Octyl 2-Cyano-3,3-diphenylacrylate
PubChem CID 22571
IUPAC Name 2-ethylhexyl 2-cyano-3,3-diphenylprop-2-enoate
SMILES CCCCC(CC)COC(=O)C(C#N)=C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point -10°C
Color Yellow
Boiling Point 218°C
Quantity 25 g
Formula Weight 361.49
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylhexyl 2-Cyano-3,3-diphenylacrylate
EINECSNumber (4)-0262
RTECSNumber UD3328750
TSCA Yes