missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylhexanal, Analytical Standard, ≥95.0% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019622
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019622 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019622 Supplier Merck Emd Millipore Supplier No. 681551ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 123-05-7
Density 0.822 g/mL (at 25°C)
Linear Formula CH3(CH2)3 CH(C2H5)CHO
MDL Number MFCD00006987
Boiling Point 55°C (13.5 mmHg)
Synonym 2-Ethylcapronaldehyde
Quantity 1 mL
Formula Weight 128.21
Percent Purity ≥95.0% (GC)
Grade Analytical Standard