missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylfuran, ≥98% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019877
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019877 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019877 Supplier Merck Emd Millipore Supplier No. 695301ML
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care

Specifications

CAS 3208-16-0
Density 0.912 g/mL (at 25°C)
Flash Point -2°C - closed cup
Physical Form Neat
Linear Formula C6H8 O
MDL Number MFCD00003259
Boiling Point 92°C to 93°C (768 mmHg)
Type Analytical Standard
Quantity 1 mL
Formula Weight 96.13
Percent Purity ≥98% (GC)
Grade Analytical Standard
Show More Show Less