missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylcyclohexanone, 99%

Catalog No. aab2074806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2074806 5 g
AAB2074814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2074806 Supplier Thermo Scientific Chemicals Supplier No. B2074806
Only null left

CAS: 4423-94-3 | C8H14O | 126.199 g/mol

2-Ethylcyclohexanone is used as a pharmaceutical intermediate. 2-Methyl and 2-ethyl derivatives of cyclohexanone were allowed to react with deuterium in tBuOD using platinum group metals as catalysts to study the substituent effects in heterogeneous catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylcyclohexanone is used as a pharmaceutical intermediate. 2-Methyl and 2-ethyl derivatives of cyclohexanone were allowed to react with deuterium in tBuOD using platinum group metals as catalysts to study the substituent effects in heterogeneous catalysis.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 4423-94-3
Molecular Formula C8H14O
Molecular Weight (g/mol) 126.199
MDL Number MFCD00066306
InChI Key WKYYYUWKFPFVEY-UHFFFAOYSA-N
Synonym 2-ethylcyclohexanone, cyclohexanone, 2-ethyl, 2-ethyl-cyclohexanone, acmc-1advf, 2-ethyl-cyclohexan-1-one
PubChem CID 20474
IUPAC Name 2-ethylcyclohexan-1-one
SMILES CCC1CCCCC1=O

Specifications

Quantity 5 g
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 126.2
Percent Purity 99%
Chemical Name or Material 2-Ethylcyclohexanone
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only