accessibility menu, dialog, popup

UserName

CAS RN 4423-94-3

IUPAC Name: 2-ethylcyclohexan-1-one
Synonyms: 2-ethylcyclohexan-1-one
Molecular Weight (g/mol): 126.2
Molecular Formula: C8H14O
InChi Key: WKYYYUWKFPFVEY-UHFFFAOYNA-N
SMILES: CCC1CCCCC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results