missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbutyric Acid 98.0+%, TCI America™

Catalog No. p-7128822
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E007025ML 25 mL
E0070500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E007025ML Supplier TCI America Supplier No. E007025ML
Only null left

Chemical Identifiers

CAS 88-09-5
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00002670
InChI Key OXQGTIUCKGYOAA-UHFFFAOYSA-N
Synonym 2-ethylbutyric acid, diethylacetic acid, butanoic acid, 2-ethyl, acetic acid, diethyl, 3-pentanecarboxylic acid, 2-ethyl butanoic acid, diethyl acetic acid, butyric acid, 2-ethyl, 2-ethyl-butyric acid, alpha-ethylbutyric acid
PubChem CID 6915
IUPAC Name 2-ethylbutanoic acid
SMILES CCC(CC)C(=O)O

Specifications

Color Colorless
Boiling Point 193°C
Quantity 25 mL
UN Number 2810
Formula Weight 116.16
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethylbutyric Acid
EINECSNumber (2)-0608
RTECSNumber ET1400000
TSCA Yes