missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbutyraldehyde 98.0+%, TCI America™

Catalog No. E0069500ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0069500ML 500 mL
E006925ML 25 mL
E0069100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0069500ML Supplier TCI America Supplier No. E0069500ML

Chemical Identifiers

CAS 97-96-1
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00006985
InChI Key UNNGUFMVYQJGTD-UHFFFAOYSA-N
Synonym 2-ethylbutyraldehyde, butanal, 2-ethyl, diethylacetaldehyde, 3-formylpentane, 2-ethylbutyric aldehyde, butyraldehyde, 2-ethyl, alpha-ethylbutanal, 2-ethylbutyric aledhyde, ethyl butyraldehyde, aldehyde 2-ethylbutyrique
PubChem CID 7359
IUPAC Name 2-ethylbutanal
SMILES CCC(CC)C=O

Specifications

Color Yellow
Boiling Point 117°C
Quantity 500 mL
UN Number 1178
Formula Weight 100.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylbutyraldehyde
EINECSNumber (2)-0494
RTECSNumber ES2625000
TSCA Yes