missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbutyl Acetate 98.0+%, TCI America™

Catalog No. p-7116669
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A002925ML 25 mL
A0029500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A002925ML Supplier TCI America Supplier No. A002925ML

Chemical Identifiers

CAS 10031-87-5
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD00009343
InChI Key HQLKZWRSOHTERR-UHFFFAOYSA-N
Synonym acetic acid, 2-ethylbutyl ester, 2-ethyl butyl acetate, beta-ethylbutyl acetate, acetic acid 2-ethylbutyl ester, 1-butanol, 2-ethyl-, acetate, fema no. 2425, 2-ethylbutylacetat, 2-ethylbutyl=acetate, acmc-2097ot, 2-ethylbutyl acetate
PubChem CID 61448
IUPAC Name 2-ethylbutyl acetate
SMILES CCC(CC)COC(=O)C

Specifications

Color Colorless
Boiling Point 159°C
Quantity 25 mL
UN Number 1177
Formula Weight 144.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethylbutyl Acetate
EINECSNumber (2)-0734&(2)-0735
TSCA Yes