accessibility menu, dialog, popup

UserName

H3CCH3OCH3O

CAS RN 10031-87-5

IUPAC Name: 2-ethylbutyl acetate
Synonyms: 1-butanol, 2-ethyl-, acetate
Molecular Weight (g/mol): 144.21
Molecular Formula: C8H16O2
InChi Key: HQLKZWRSOHTERR-UHFFFAOYSA-N
SMILES: CCC(CC)COC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results