missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylbenzaldehyde, 97%

Catalog No. AC460550050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460550050 5 g
AC460550250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460550050 Supplier Thermo Scientific Chemicals Supplier No. 460550050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22927-13-5 | C9H10O | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22927-13-5
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD02261766
InChI Key NTWBHJYRDKBGBR-UHFFFAOYSA-N
Synonym benzaldehyde, 2-ethyl, o-ethylbenzaldehyde, 2-ethyl benzaldehyde, 2-ethyl-benzaldehyde, ethylbenzaldehyde, 2-ethyl-benzaldehyd, 6-ethyl benzaldehyde, pubchem3074, acmc-20a49p
PubChem CID 123406
IUPAC Name 2-ethylbenzaldehyde
SMILES CCC1=CC=CC=C1C=O

Specifications

Density 1.0200g/mL
Boiling Point 209.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Refractive Index 1.538
Quantity 5 g
Specific Gravity 1.02
Formula Weight 134.18
Percent Purity 97%
Chemical Name or Material 2-Ethylbenzaldehyde
Show More Show Less

RUO – Research Use Only