missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylaniline 98.0+%, TCI America™

Catalog No. p-7128814
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mL
500 mL
Catalog No. Quantity
E006025ML 25 mL
E0060500ML 500 mL
2 options

Catalog No. E006025ML

Supplier: TCI America E006025ML

Only null left

Chemical Identifiers

CAS 578-54-1
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00007751
InChI Key MLPVBIWIRCKMJV-UHFFFAOYSA-N
Synonym o-ethylaniline, benzenamine, 2-ethyl, aniline, o-ethyl, 2-ethylbenzenamine, o-aminoethylbenzene, aniline, 2-ethyl, 2-ethyl aniline, 2-ethylphenylamine, unii-gr557n47k6, ccris 2858
PubChem CID 11357
IUPAC Name 2-ethylaniline
SMILES CCC1=CC=CC=C1N

Specifications

Melting Point -44°C
Color Yellow
Boiling Point 216°C
Quantity 25 mL
UN Number 2273
Formula Weight 121.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethylaniline
EINECSNumber (3)-0206
RTECSNumber BX9800000
TSCA Yes