Learn More
2-Ethylaniline, 97%
CAS: 578-54-1 | C8H11N | 121.183 g/mol
$48.19 - $48.19
Chemical Identifiers
| CAS | 578-54-1 |
|---|---|
| Molecular Formula | C8H11N |
| Molecular Weight (g/mol) | 121.183 |
| MDL Number | MFCD00007751 |
| InChI Key | MLPVBIWIRCKMJV-UHFFFAOYSA-N |
| Synonym | o-ethylaniline, benzenamine, 2-ethyl, aniline, o-ethyl, 2-ethylbenzenamine, o-aminoethylbenzene, aniline, 2-ethyl, 2-ethyl aniline, 2-ethylphenylamine, unii-gr557n47k6, ccris 2858 |
| PubChem CID | 11357 |
| IUPAC Name | 2-ethylaniline |
| SMILES | CCC1=CC=CC=C1N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1498422
|
Thermo Scientific Chemicals
A1498422 |
100 g |
Each for $48.19
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 578-54-1 | |
| 121.183 | |
| MLPVBIWIRCKMJV-UHFFFAOYSA-N | |
| 11357 | |
| CCC1=CC=CC=C1N |
| C8H11N | |
| MFCD00007751 | |
| o-ethylaniline, benzenamine, 2-ethyl, aniline, o-ethyl, 2-ethylbenzenamine, o-aminoethylbenzene, aniline, 2-ethyl, 2-ethyl aniline, 2-ethylphenylamine, unii-gr557n47k6, ccris 2858 | |
| 2-ethylaniline |
Specifications
| 578-54-1 | |
| 0.983 | |
| 91°C (196°F) | |
| C8H11N | |
| MFCD00007751 | |
| UN2273 | |
| o-ethylaniline, benzenamine, 2-ethyl, aniline, o-ethyl, 2-ethylbenzenamine, o-aminoethylbenzene, aniline, 2-ethyl, 2-ethyl aniline, 2-ethylphenylamine, unii-gr557n47k6, ccris 2858 | |
| CCC1=CC=CC=C1N | |
| 121.183 | |
| 121.18 | |
| 2-Ethylaniline |
| -47°C | |
| 208°C to 210°C | |
| Characteristic | |
| 1.56 | |
| 100 g | |
| 1841268 | |
| MLPVBIWIRCKMJV-UHFFFAOYSA-N | |
| 2-ethylaniline | |
| 11357 | |
| 97% |
Safety and Handling
GHS H Statement
H331-H373-H227-H302-H312
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
P210-P233-P240-P243-P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P362-P374-P501c
H227-H301+H311+H331-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: 2-ETHYLANILINE
EINECSNumber : 209-424-2
RTECSNumber : BX9800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only