missing translation for 'onlineSavingsMsg'
Learn More

2-(Ethylamino)ethanol, 98%

Catalog No. p-126855
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 kg
250 g
Packaging:
Glass bottle
Catalog No. Quantity Packaging
AC168732500 250 g Glass bottle
AC168730010 1 kg Glass bottle
2 options

Catalog No. AC168732500

Supplier: Thermo Scientific Chemicals 168732500

Only null left

CAS: 110-73-6 | C4H11NO | 89.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-73-6
Molecular Formula C4H11NO
Molecular Weight (g/mol) 89.14
MDL Number MFCD00002841
InChI Key MIJDSYMOBYNHOT-UHFFFAOYSA-N
Synonym 2-ethylamino ethanol, ethanol, 2-ethylamino, ethyl ethanolamine, n-ethylethanolamine, 2-hydroxyethyl ethylamine, n-ethylmonoethanolamine, 2-n-monoethylaminoethanol, n-ethylaminoethanol, n-ethyl ethanolamine, 2-ethylamino ethan-1-ol
PubChem CID 8072
IUPAC Name 2-(ethylamino)ethanol
SMILES CCNCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -6°C
Color Colorless to Yellow
Density 0.9100g/mL
Boiling Point 167°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4390 to 1.441
Linear Formula C2H5NHCH2CH2OH
Quantity 250 g
Beilstein 04, 282
Specific Gravity 0.91
Solubility Information Solubility in water: miscible. Other solubilities: soluble in alcohol and ether
Formula Weight 89.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(Ethylamino)ethanol, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
EINECSNumber 203-797-5
RTECSNumber KK9100000
TSCA TSCA

RUO – Research Use Only