Learn More
2-Ethylacrolein, tech. 90%, stab. with 50ppm hydroquinone
CAS: 922-63-4 | C5H8O | 84.12 g/mol
Supplier: Thermo Scientific Chemicals B2183914
Description
2-Ethylacrolein has been used in total synthesis of (±)-tabersonine, Aspidosperma alkaloid and synthesis of conjugate with bovine serum albumin tethered to Tn.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Ethylacrolein has been used in total synthesis of (+/-)-tabersonine, Aspidosperma alkaloid and synthesis of conjugate with bovine serum albumin tethered to Tn.
Solubility
Soluble in alcohol. Insoluble in water.
Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store at 2-8°C.
Specifications
| 2-Ethylacrolein | |
| 922-63-4 | |
| 92°C to 93°C | |
| Pungent | |
| 1.4245 | |
| 25 g | |
| 1098378 | |
| 2-ethylacrolein, 2-methylenebutanal, 2-ethylpropenal, butanal, 2-methylene, ethacrolein, 2-ethylacrylaldehyde, 2-methylenebutyraldehyde, ethylacrolein, alpha-ethylacrolein, alpha-ethylacrylaldehyde | |
| GMLDCZYTIPCVMO-UHFFFAOYSA-N | |
| 2-methylidenebutanal | |
| 70203 | |
| 90% |
| stabilized with 50ppm hydroquinone | |
| 0.859 | |
| 1°C (34°F) | |
| C5H8O | |
| MFCD00010129 | |
| UN1992 | |
| Air sensitive | |
| Soluble in alcohol. Insoluble in water. | |
| CCC(=C)C=O | |
| 84.12 | |
| 84.12 | |
| Technical |
Chemical Identifiers
| 922-63-4 | |
| 84.12 | |
| GMLDCZYTIPCVMO-UHFFFAOYSA-N | |
| 70203 | |
| CCC(=C)C=O |
| C5H8O | |
| MFCD00010129 | |
| 2-ethylacrolein, 2-methylenebutanal, 2-ethylpropenal, butanal, 2-methylene, ethacrolein, 2-ethylacrylaldehyde, 2-methylenebutyraldehyde, ethylacrolein, alpha-ethylacrolein, alpha-ethylacrylaldehyde | |
| 2-methylidenebutanal |
Safety and Handling
GHS H Statement
H225-H330-H341-H315-H319-H335
Highly flammable liquid and vapour.
Fatal if inhaled.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P312-P303+P361+P353-P304+P340-P310-P312-P330-P363-P370+P378q-P501c
H225-H302+H312-H330
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber : 213-079-3
RTECSNumber : ES3480000
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only