missing translation for 'onlineSavingsMsg'
Learn More

2-Ethylacrolein, tech. 90%, stab. with 50ppm hydroquinone

Catalog No. p-7026057
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2183906 5 g
AAB2183914 25 g
AAB2183922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2183906 Supplier Thermo Scientific Chemicals Supplier No. B2183906

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 922-63-4 | C5H8O | 84.12 g/mol

2-Ethylacrolein has been used in total synthesis of (±)-tabersonine, Aspidosperma alkaloid and synthesis of conjugate with bovine serum albumin tethered to Tn.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylacrolein has been used in total synthesis of (+/-)-tabersonine, Aspidosperma alkaloid and synthesis of conjugate with bovine serum albumin tethered to Tn.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Air sensitive. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store at 2-8°C.

Chemical Identifiers

CAS 922-63-4
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00010129
InChI Key GMLDCZYTIPCVMO-UHFFFAOYSA-N
Synonym 2-ethylacrolein, 2-methylenebutanal, 2-ethylpropenal, butanal, 2-methylene, ethacrolein, 2-ethylacrylaldehyde, 2-methylenebutyraldehyde, ethylacrolein, alpha-ethylacrolein, alpha-ethylacrylaldehyde
PubChem CID 70203
IUPAC Name 2-methylidenebutanal
SMILES CCC(=C)C=O

Specifications

Density 0.859
Boiling Point 92°C to 93°C
Flash Point 1°C (34°F)
Odor Pungent
Refractive Index 1.4245
Quantity 5 g
UN Number UN1992
Beilstein 1098378
Sensitivity Air sensitive
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 84.12
Percent Purity 90%
Grade Technical
Chemical Name or Material 2-Ethylacrolein, stabilized with 50ppm hydroquinone
Show More Show Less
Hazard Category H225-H302+H312-H330
Hazard Statement GHS H Statement
H225-H330-H341-H315-H319-H335
Highly flammable liquid and vapour.
Fatal if inhaled.
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P312-P303+P361+P353-P304+P340-P310-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 213-079-3
RTECSNumber ES3480000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only