missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-6-methylphenol 98.0+%, TCI America™

Catalog No. E11851G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11851G 1 g
E11855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E11851G Supplier TCI America Supplier No. E11851G

Chemical Identifiers

CAS 1687-64-5
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00962292
InChI Key CIRRFAQIWQFQSS-UHFFFAOYSA-N
Synonym phenol, 2-ethyl-6-methyl, o-cresol, 6-ethyl, 6-ethyl-o-cresol, unii-359wuc62ri, 2-ethyl-6-methyl-phenol, 6-ethyl-2-methylphenol, acmc-20anwj, 2-methyl-6-ethylphenol, 2-ethyl-6-methyl phenol, phenol,2-ethyl-6-methyl
PubChem CID 519333
IUPAC Name 2-ethyl-6-methylphenol
SMILES CCC1=CC=CC(=C1O)C

Specifications

Boiling Point 214°C
Quantity 1 g
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-6-methylphenol
EINECSNumber (3)-0521&(4)-0057
TSCA No