missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-5,5-dimethyl-1,3-dioxane solution in acetonitrile (1000mg/L), Thermo Scientific™

Catalog No. AAH32308AE
Change view
Click to view available options
Quantity:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32308AE 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH32308AE Supplier Thermo Scientific Chemicals Supplier No. H32308AE

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

 

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 768-58-1
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD16622268
InChI Key QSHOOPIYPOINNH-UHFFFAOYSA-N
Synonym solution in acetonitrile 1000mg/l, 1,3-dioxane, 2-ethyl-5,5-dimethyl
PubChem CID 69852
IUPAC Name 2-ethyl-5,5-dimethyl-1,3-dioxane
SMILES CCC1OCC(CO1)(C)C

Specifications

Quantity 100mL
UN Number UN1648
Concentration or Composition (by Analyte or Components) 1000mg/L
Chemical Name or Material 2-Ethyl-5,5-dimethyl-1,3-dioxane solution in acetonitrile
Hazard Category H225-H302+H312+H332-H319
Hazard Statement GHS H Statement
H225-H312-H332-H319
Highly flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONITRILE
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only