CAS RN 768-58-1
IUPAC Name:
2-ethyl-5,5-dimethyl-1,3-dioxane
Synonyms:
2-ethyl-5,5-dimethyl-1,3-dioxane
Molecular Weight (g/mol):
144.21
Molecular Formula:
C8H16O2
InChi Key:
QSHOOPIYPOINNH-UHFFFAOYSA-N
SMILES:
CCC1OCC(C)(C)CO1
1
–
2
of
2
results
1
Promotions Available
2-Ethyl-5,5-dimethyl-1,3-dioxane, 99%
2-Ethyl-5,5-dimethyl-1,3-dioxane, 99%
CAS: 768-58-1 Molecular Formula: C8H16O2 Molecular Weight (g/mol): 144.214 MDL Number: MFCD16622268 InChI Key: QSHOOPIYPOINNH-UHFFFAOYSA-N Synonym: solution in acetonitrile 1000mg/l,1,3-dioxane, 2-ethyl-5,5-dimethyl PubChem CID: 69852 IUPAC Name: 2-ethyl-5,5-dimethyl-1,3-dioxane SMILES: CCC1OCC(CO1)(C)C
2
Promotions Available
2-Ethyl-5,5-dimethyl-1,3-dioxane solution in acetonitrile (1000mg/L), Thermo Scientific™
2-Ethyl-5,5-dimethyl-1,3-dioxane solution in acetonitrile (1000mg/L), Thermo Scientific™
CAS: 768-58-1 Molecular Formula: C8H16O2 Molecular Weight (g/mol): 144.214 MDL Number: MFCD16622268 InChI Key: QSHOOPIYPOINNH-UHFFFAOYSA-N Synonym: solution in acetonitrile 1000mg/l,1,3-dioxane, 2-ethyl-5,5-dimethyl PubChem CID: 69852 IUPAC Name: 2-ethyl-5,5-dimethyl-1,3-dioxane SMILES: CCC1OCC(CO1)(C)C