missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-5,5-dimethyl-1,3-dioxane, 99%

Catalog No. AAH3235106
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3235106 5 g
AAH3235114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3235106 Supplier Thermo Scientific Chemicals Supplier No. H3235106
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 768-58-1 | C8H16O2 | 144.214 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 768-58-1
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.214
MDL Number MFCD16622268
InChI Key QSHOOPIYPOINNH-UHFFFAOYSA-N
Synonym solution in acetonitrile 1000mg/l, 1,3-dioxane, 2-ethyl-5,5-dimethyl
PubChem CID 69852
IUPAC Name 2-ethyl-5,5-dimethyl-1,3-dioxane
SMILES CCC1OCC(CO1)(C)C

Specifications

Density 0.88
Boiling Point 154°C
Flash Point 50°C (122°F)
Odor Characteristic
Quantity 5 g
UN Number UN1993
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material 2-Ethyl-5,5-dimethyl-1,3-dioxane
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only