missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethyl-4-methylimidazole 95.0+%, TCI America™
Supplier: TCI America E0232100G
Specifications
| 2-Ethyl-4-methylimidazole | |
| 40°C | |
| 164°C | |
| MFCD00005193 | |
| 2-ethyl-4-methylimidazole, 2-ethyl-4-methyl-1h-imidazole, 1h-imidazole, 2-ethyl-4-methyl, 4-methyl-2-ethylimidazole, 1h-imidazole, 2-ethyl-5-methyl, imidazole, 2-ethyl-4-methyl, unii-5k8xi641g3, 2-ethyl-4-methyl-imidazole, 2-ethyl-4-methyl-3h-imidazole, curazol 2e4mz | |
| CCC1=NC=C(N1)C | |
| 110.16 | |
| 110.16 | |
| Crystalline Lumps |
| 931-36-2 | |
| White-Yellow | |
| C6H10N2 | |
| 100 g | |
| ULKLGIFJWFIQFF-UHFFFAOYSA-N | |
| 2-ethyl-5-methyl-1H-imidazole | |
| 70262 | |
| ≥95.0% (GC) |
Chemical Identifiers
| 931-36-2 | |
| 110.16 | |
| ULKLGIFJWFIQFF-UHFFFAOYSA-N | |
| 70262 | |
| CCC1=NC=C(N1)C |
| C6H10N2 | |
| MFCD00005193 | |
| 2-ethyl-4-methylimidazole, 2-ethyl-4-methyl-1h-imidazole, 1h-imidazole, 2-ethyl-4-methyl, 4-methyl-2-ethylimidazole, 1h-imidazole, 2-ethyl-5-methyl, imidazole, 2-ethyl-4-methyl, unii-5k8xi641g3, 2-ethyl-4-methyl-imidazole, 2-ethyl-4-methyl-3h-imidazole, curazol 2e4mz | |
| 2-ethyl-5-methyl-1H-imidazole |
Safety and Handling
EINECSNumber : (5)-0388
RTECSNumber : NI6147500
TSCA : Yes