accessibility menu, dialog, popup

UserName

H3CNHCH3N

CAS RN 931-36-2

IUPAC Name: 2-ethyl-5-methyl-1H-imidazole
Synonyms: 2-ethyl-4-methyl-3H-imidazole
Molecular Weight (g/mol): 110.16
Molecular Formula: C6H10N2
InChi Key: ULKLGIFJWFIQFF-UHFFFAOYSA-N
SMILES: CCC1=NC=C(C)N1
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (3)