missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-4-methylimidazole 95.0+%, TCI America™

Catalog No. E0232500G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0232500G 500 g
E023225G 25 g
E0232100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E0232500G Supplier TCI America Supplier No. E0232500G

Chemical Identifiers

CAS 931-36-2
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD00005193
InChI Key ULKLGIFJWFIQFF-UHFFFAOYSA-N
Synonym 2-ethyl-4-methylimidazole, 2-ethyl-4-methyl-1h-imidazole, 1h-imidazole, 2-ethyl-4-methyl, 4-methyl-2-ethylimidazole, 1h-imidazole, 2-ethyl-5-methyl, imidazole, 2-ethyl-4-methyl, unii-5k8xi641g3, 2-ethyl-4-methyl-imidazole, 2-ethyl-4-methyl-3h-imidazole, curazol 2e4mz
PubChem CID 70262
IUPAC Name 2-ethyl-5-methyl-1H-imidazole
SMILES CCC1=NC=C(N1)C

Specifications

Melting Point 40°C
Color White-Yellow
Boiling Point 164°C
Quantity 500 g
Formula Weight 110.16
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2-Ethyl-4-methylimidazole
EINECSNumber (5)-0388
RTECSNumber NI6147500
TSCA Yes