missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-4-methylimidazole, 96%

Catalog No. p-7022850
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1579814 25 g
AAA1579822 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1579814 Supplier Thermo Scientific Chemicals Supplier No. A1579814
Only null left

CAS: 931-36-2 | C6H10N2 | 110.16 g/mol

2-Ethyl-4-methyl-1H-imidazole is a useful synthetic intermediate. It can be used as a building block for active ingredients as well as in epoxy curing. It is used as an intermediate for pharmaceuticals, agrochemicals, dyes, textile auxiliaries, pigments and other organic chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethyl-4-methyl-1H-imidazole is a useful synthetic intermediate. It can be used as a building block for active ingredients as well as in epoxy curing. It is used as an intermediate for pharmaceuticals, agrochemicals, dyes, textile auxiliaries, pigments and other organic chemicals.

Solubility
Soluble in water (<210 g/1000 ml at 20°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 931-36-2
Molecular Formula C6H10N2
Molecular Weight (g/mol) 110.16
MDL Number MFCD00005193
InChI Key ULKLGIFJWFIQFF-UHFFFAOYSA-N
Synonym 2-ethyl-4-methylimidazole, 2-ethyl-4-methyl-1h-imidazole, 1h-imidazole, 2-ethyl-4-methyl, 4-methyl-2-ethylimidazole, 1h-imidazole, 2-ethyl-5-methyl, imidazole, 2-ethyl-4-methyl, unii-5k8xi641g3, 2-ethyl-4-methyl-imidazole, 2-ethyl-4-methyl-3h-imidazole, curazol 2e4mz
PubChem CID 70262
IUPAC Name 2-ethyl-5-methyl-1H-imidazole
SMILES CCC1=NC=C(N1)C

Specifications

Melting Point 45°C to 49°C
Density 0.973
Boiling Point 292°C to 295°C
Flash Point 137°C (278°F)
Refractive Index 1.5 00
Quantity 25 g
Beilstein 1711
Solubility Information Soluble in water (< 210g/1000 ml at 20°C).
Formula Weight 110.16
Percent Purity 96%
Chemical Name or Material 2-Ethyl-4-methylimidazole
Show More Show Less
Hazard Category H302-H315-H317-H318-H335-H351
Hazard Statement GHS H Statement
H318-H302-H335-H315
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P310-P330-P333+P313-P362-P501c
EINECSNumber 213-234-5
RTECSNumber NI6147500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only