missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-3,5(6)-dimethylpyrazine, 99%, mixture of isomers

Catalog No. p-7029869
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2738306 5 g
AAH2738314 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2738306 Supplier Thermo Scientific Chemicals Supplier No. H2738306
Only null left

CAS: 13925-07-0 | C8H12N2 | 136.198 g/mol

It is applied as an odorant of coffee brews, nutty flavor products and roasted food products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is applied as an odorant of coffee brews, nutty flavor products and roasted food products.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol, chloroform and dichloromethane.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 13925-07-0
Molecular Formula C8H12N2
Molecular Weight (g/mol) 136.198
MDL Number MFCD00047392
InChI Key JZBCTZLGKSYRSF-UHFFFAOYSA-N
Synonym 3,5-dimethyl-2-ethylpyrazine, pyrazine, 2-ethyl-3,5-dimethyl, 2,6-dimethyl-3-ethylpyrazine, 3-ethyl-2,6-dimethylpyrazine, 2-ethyl-3,5-dimethyl pyrazine, pyrazine, 2,6-dimethyl-3-ethyl, unii-r182ey5l8c, fema no. 3149, 2-ethyl-3,5 6-dimethylpyrazine, pyrazine, 3,5-dimethyl-2-ethyl
PubChem CID 26334
IUPAC Name 2-ethyl-3,5-dimethylpyrazine
SMILES CCC1=NC=C(N=C1C)C

Specifications

Density 0.96
Boiling Point 182°C
Flash Point 66°C (151°F)
Refractive Index 1.5015
Quantity 5 g
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol,chloroform and dichloromethane.
Formula Weight 136.19
Percent Purity 99%
Chemical Name or Material 2-Ethyl-3,5(6)-dimethylpyrazine, mixture of isomers
Hazard Category H227-H302
Hazard Statement GHS H Statement
H302-H227
Harmful if swallowed.
Combustible liquid.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
EINECSNumber 237-694-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only