missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 164650025

 View more versions of this product

Catalog No. AC164650025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2-Ethyl-2-(hydroxymethyl)-1, 3-propanediol
0.002% max. (as formic acid)
100.0
172°C
98%
C6H14O3
MFCD00004694
01, III, 2349
Solubility in water: soluble. Other solubilities: freely soluble in alcohols,insoluble in hydrocarbons
CCC(CO)(CO)CO
134.18
157 mPa.s (75°C)
98%
77-99-6
97.5
160°C (0.7 hPa)
Authentic
Plastic bottle
C2H5C(CH2OH)3
2.5kg
trimethylolpropane, 2-ethyl-2-hydroxymethyl propane-1,3-diol, ethriol, 2-ethyl-2-hydroxymethyl-1,3-propanediol, hexaglycerine, ettriol, etriol, trimethylol propane, tmp alcohol, 1,1,1-tris hydroxymethyl propane
ZJCCRDAZUWHFQH-UHFFFAOYSA-N
2-ethyl-2-(hydroxymethyl)propane-1,3-diol
6510
134.18

Chemical Identifiers

77-99-6
134.18
ZJCCRDAZUWHFQH-UHFFFAOYSA-N
6510
CCC(CO)(CO)CO
C6H14O3
MFCD00004694
trimethylolpropane, 2-ethyl-2-hydroxymethyl propane-1,3-diol, ethriol, 2-ethyl-2-hydroxymethyl-1,3-propanediol, hexaglycerine, ettriol, etriol, trimethylol propane, tmp alcohol, 1,1,1-tris hydroxymethyl propane
2-ethyl-2-(hydroxymethyl)propane-1,3-diol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 201-074-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only