missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-2-(hydroxymethyl)-1,3-propanediol, 98%

Catalog No. p-3736450
Change view
Click to view available options
Quantity:
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC164655000 500 g Plastic bottle
AC164650010 1 kg Plastic bottle
AC164650025 2.5 kg Plastic bottle
AC164650100 10 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC164655000 Supplier Thermo Scientific Chemicals Supplier No. 164655000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 77-99-6 | C6H14O3 | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77-99-6
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00004694
InChI Key ZJCCRDAZUWHFQH-UHFFFAOYSA-N
Synonym trimethylolpropane, 2-ethyl-2-hydroxymethyl propane-1,3-diol, ethriol, 2-ethyl-2-hydroxymethyl-1,3-propanediol, hexaglycerine, ettriol, etriol, trimethylol propane, tmp alcohol, 1,1,1-tris hydroxymethyl propane
PubChem CID 6510
IUPAC Name 2-ethyl-2-(hydroxymethyl)propane-1,3-diol
SMILES CCC(CO)(CO)CO

Specifications

Acidity 0.002% max. (as formic acid)
CAS Min % 97.5
CAS Max % 100.0
Melting Point 56.0°C to 60.0°C
Color White
Boiling Point 160°C (0.7 hPa)
Flash Point 172°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Linear Formula C2H5C(CH2OH)3
Quantity 500 g
Beilstein 01, III, 2349
Solubility Information Solubility in water: soluble. Other solubilities: freely soluble in alcohols,insoluble in hydrocarbons
Viscosity 157 mPa.s (75°C)
Formula Weight 134.18
Percent Purity 98%
Physical Form Flakes
Chemical Name or Material 2-Ethyl-2-(hydroxymethyl)-1, 3-propanediol
Show More Show Less
EINECSNumber 201-074-9

RUO – Research Use Only