missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1-butanol 98.0+%, TCI America™

Catalog No. p-7128820
Change view
Click to view available options
Quantity:
100 mL
25 mL
500 mL
Catalog No. Quantity
E006825ML 25 mL
E0068100ML 100 mL
E0068500ML 500 mL
3 options

Catalog No. E006825ML

Supplier: TCI America E006825ML

Only null left

Chemical Identifiers

CAS 97-95-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004744
InChI Key TZYRSLHNPKPEFV-UHFFFAOYSA-N
Synonym 2-ethyl-1-butanol, 1-butanol, 2-ethyl, 2-ethylbutanol, 2-ethylbutyl alcohol, pseudohexyl alcohol, 3-methylolpentane, 2-ethylbutanol-1, ethylbutanol, 3-pentylcarbinol, sec-pentylcarbinol
PubChem CID 7358
IUPAC Name 2-ethylbutan-1-ol
SMILES CCC(CC)CO

Specifications

Melting Point -15°C
Color Colorless
Boiling Point 146°C
Quantity 25 mL
UN Number 2275
Formula Weight 102.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-1-butanol
EINECSNumber (2)-0217
RTECSNumber EL3850000
TSCA Yes