accessibility menu, dialog, popup

UserName

CH3H3CHO

CAS RN 97-95-0

IUPAC Name: 2-ethylbutan-1-ol
Synonyms: 2-ethylbutanol
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: TZYRSLHNPKPEFV-UHFFFAOYSA-N
SMILES: CCC(CC)CO
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)