missing translation for 'onlineSavingsMsg'
Learn More

2-Ethyl-1-butanol 98.0+%, TCI America™

Catalog No. E006825ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E006825ML 25 mL
E0068100ML 100 mL
E0068500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E006825ML Supplier TCI America Supplier No. E006825ML

Chemical Identifiers

CAS 97-95-0
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004744
InChI Key TZYRSLHNPKPEFV-UHFFFAOYSA-N
Synonym 2-ethyl-1-butanol, 1-butanol, 2-ethyl, 2-ethylbutanol, 2-ethylbutyl alcohol, pseudohexyl alcohol, 3-methylolpentane, 2-ethylbutanol-1, ethylbutanol, 3-pentylcarbinol, sec-pentylcarbinol
PubChem CID 7358
IUPAC Name 2-ethylbutan-1-ol
SMILES CCC(CC)CO

Specifications

Melting Point -15°C
Color Colorless
Boiling Point 146°C
Quantity 25 mL
UN Number 2275
Formula Weight 102.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethyl-1-butanol
EINECSNumber (2)-0217
RTECSNumber EL3850000
TSCA Yes