missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxypyridine-5-boronic acid pinacol ester, 99%, Thermo Scientific Chemicals

Catalog No. p-7033125
Change view
Click to view available options
Quantity:
1 g
Catalog No. Quantity
AAH5013703 1 g
1 options

Catalog No. AAH5013703

Supplier: Thermo Scientific Chemicals H5013703

Only null left

2-Ethoxypyridine-5-boronic acid pinacol ester is used as raw material for synthesis of pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxypyridine-5-boronic acid pinacol ester is used as raw material for synthesis of pharmaceutical intermediates.

Solubility
Sparingly soluble in water (0.019 g/L) (25°C)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 1072945-01-7
Molecular Formula C13H20BNO3
Molecular Weight (g/mol) 249.117
MDL Number MFCD07781182
InChI Key GXUHWEZZHFBDFA-UHFFFAOYSA-N
Synonym 2-ethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 6-ethoxypyridine-3-boronic acid pinacol ester, 2-ethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-ethoxypyridine-5-boronic acid pinacol ester, 2-ethoxypyridine-5-boronic acid, pinacol ester, pubchem20146, 6-ethoxypyridine-3-boronicacidpinacolester, 6-ethoxypyridin-3-ylboronic acid pinacol ester, 2-ethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-ethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine
PubChem CID 24208796
IUPAC Name 2-ethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCC

Specifications

Quantity 1 g
Solubility Information Sparingly soluble in water (0.019g/L) (25°C)
Formula Weight 249.12
Percent Purity 99%
Chemical Name or Material 2-Ethoxypyridine-5-boronic acid pinacol ester
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only