missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethyl Acetate 98.0+%, TCI America™

Catalog No. p-7128807
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E004925ML 25 mL
E0049500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E004925ML Supplier TCI America Supplier No. E004925ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 111-15-9
Molecular Formula C6H12O3
Molecular Weight (g/mol) 132.16
MDL Number MFCD00009251
InChI Key SVONRAPFKPVNKG-UHFFFAOYSA-N
Synonym ethylene glycol monoethyl ether acetate, cellosolve acetate, oxitol acetate, ethoxyethyl acetate, oxytol acetate, ethylglycol acetate, ethyl cellosolve acetate, 1-acetoxy-2-ethoxyethane, ethylglykolacetat, octan etoksyetylu
PubChem CID 8095
IUPAC Name 2-ethoxyethyl acetate
SMILES CCOCCOC(C)=O

Specifications

Melting Point -62°C
Color Colorless
Boiling Point 156°C
Quantity 25 mL
UN Number 1172
Formula Weight 132.16
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethyl Acetate
EINECSNumber (2)-0740
RTECSNumber KK8225000
TSCA Yes