missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethyl acetate, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A13639AP

 View more versions of this product

Catalog No. AAA13639AP


Only null left
Explore available promotions

Description

Description

2-Ethoxyethyl acetate is used as a solvent dissolve resin, leather, ink, for the formulation of paints, lacquers and varnishes for industrial use. It is also used to study the field evaluation of a passive badge for measuring 2-ethoxyethyl acetate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxyethyl acetate is used as a solvent dissolve resin, leather, ink, for the formulation of paints, lacquers and varnishes for industrial use. It is also used to study the field evaluation of a passive badge for measuring 2-ethoxyethyl acetate.

Solubility
Soluble in water(230g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2-Ethoxyethyl acetate
-62°C
155°C to 157°C
Fruit-like
1.407
MFCD00009251
UN1172
14,3751
Soluble in water(230g/L).
CCOCCOC(C)=O
132.16
132.16
111-15-9
0.975
52°C (126°F)
C6H12O3
CH3CO2CH2CH2OCH2CH3
500 mL
1748677
ethylene glycol monoethyl ether acetate, cellosolve acetate, oxitol acetate, ethoxyethyl acetate, oxytol acetate, ethylglycol acetate, ethyl cellosolve acetate, 1-acetoxy-2-ethoxyethane, ethylglykolacetat, octan etoksyetylu
SVONRAPFKPVNKG-UHFFFAOYSA-N
2-ethoxyethyl acetate
8095
≥98%

Chemical Identifiers

111-15-9
132.16
SVONRAPFKPVNKG-UHFFFAOYSA-N
8095
CCOCCOC(C)=O
C6H12O3
MFCD00009251
ethylene glycol monoethyl ether acetate, cellosolve acetate, oxitol acetate, ethoxyethyl acetate, oxytol acetate, ethylglycol acetate, ethyl cellosolve acetate, 1-acetoxy-2-ethoxyethane, ethylglykolacetat, octan etoksyetylu
2-ethoxyethyl acetate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.

P201-P202-P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P308+P313-P312-P330-P363-P374-P380-P501c

H226-H302+H312+H332-H360FD

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOETHYL ETHER ACETATE

EINECSNumber : 203-839-2

RTECSNumber : KK8225000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only