missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethanol 99.0+%, TCI America™

Catalog No. p-7128805
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E004725ML 25 mL
E0047500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E004725ML Supplier TCI America Supplier No. E004725ML
Only null left

Chemical Identifiers

CAS 110-80-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002869
InChI Key ZNQVEEAIQZEUHB-UHFFFAOYSA-N
Synonym cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
PubChem CID 8076
ChEBI CHEBI:46788
IUPAC Name 2-ethoxyethan-1-ol
SMILES CCOCCO

Specifications

Color Colorless
Boiling Point 135°C
Quantity 25 mL
UN Number 1171
Formula Weight 90.12
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-Ethoxyethanol
EINECSNumber (2)-0411&(2)-2424&(7)-0097
RTECSNumber KK8050000
TSCA Yes