missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxyethanol, Spectrum™ Chemical
Specifications
Specifications
| Acidity | 5e-05 |
| CAS Max % | 1 |
| Boiling Point | 135.1°C (275.2°F) |
| Packaging | Amber Glass Bottle |
| Quantity | 200 L |
| Specific Gravity | 0.9290 - 0.9330 |
| Grade | Reagent |
| Physical Form | Liquid |
| Chemical Name or Material | 2-Ethoxyethanol |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.