missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxyethanol, Reagent Grade, 99%, Solstice
Description
- ≥95% for general laboratory use.
Specifications
Specifications
| CAS Max % | 100.0000% |
| Melting Point | -90°C (literature) |
| Density | 0.93 g/mL at 25 °C(lit.) |
| Boiling Point | 136°C |
| Flash Point | 40°C |
| Packaging | Glass Bottle |
| Linear Formula | FeCl3 |
| Quantity | 1 L |
| UN Number | UN1171 |
| Beilstein | 1098271 |
| Show More |
Safety and Handling
| ShelfLife | 1080 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Ethylene glycol monoethyl ether |
| EINECSNumber | 203-804-1 |
| Recommended Storage | Room Temp |