missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethanol, Reagent Grade, 99%, Solstice

Catalog No. 60044757
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60044757 1 L Glass Bottle
60044758 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60044757 Supplier Solstice Supplier No. 1280821L

Reagent Grade, 99%

  • ≥95% for general laboratory use.

Chemical Identifiers

CAS 110-80-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002869
InChI Key ZNQVEEAIQZEUHB-UHFFFAOYSA-N
Synonym cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
PubChem CID 8076
ChEBI CHEBI:46788
SMILES CCOCCO

Specifications

CAS Max % 100.0000%
Melting Point -90°C (literature)
Density 0.93 g/mL at 25 °C(lit.)
Boiling Point 136°C
Flash Point 40°C
Packaging Glass Bottle
Linear Formula FeCl3
Quantity 1 L
UN Number UN1171
Beilstein 1098271
IUPAC Name 2-ethoxyethanol
Formula Weight 90.12
Percent Purity >98.5%
Grade Reagent
Chemical Name or Material 2-Ethoxyethanol, Reagent Grade, 99%
Show More Show Less
ShelfLife 1080 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: III; Proper Shipping Name: Ethylene glycol monoethyl ether
EINECSNumber 203-804-1
Recommended Storage Room Temp