missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxyethanol, for analysis

Catalog No. AC445760010
Change view
Click to view available options
Quantity:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC445760010 1 L
AC445760025 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC445760010 Supplier Thermo Scientific Chemicals Supplier No. 445760010

CAS: 110-80-5 | C4H10O2 | 90.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 110-80-5
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.12
MDL Number MFCD00002869
InChI Key ZNQVEEAIQZEUHB-UHFFFAOYSA-N
Synonym cellosolve, oxitol, ethyl cellosolve, ethylene glycol monoethyl ether, ethanol, 2-ethoxy, ethyl glycol, emkanol, hydroxy ether, ethylene glycol ethyl ether, dowanol ee
PubChem CID 8076
ChEBI CHEBI:46788
IUPAC Name 2-ethoxyethanol
SMILES CCOCCO

Specifications

Melting Point -70°C
Density 0.9300g/mL
Boiling Point 135.0°C
Flash Point 40°C
Infrared Spectrum Authentic
Refractive Index 1.4060 to 1.4100
Linear Formula C2H5OCH2CH2OH
Quantity 1 L
Residue after Evaporation 0.005% max.
Merck Index 15,3803
Specific Gravity 0.93
Solubility Information Solubility in water: complete solubility. Other solubilities: miscible with most organic solvents
Viscosity 2.08 mPa.s (20°C)
Formula Weight 90.12
Percent Purity ≥98.5% (GC)
Grade Analysis
Chemical Name or Material 2-Ethoxyethanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Flammable liquid and vapour.
May damage fertility.
May damage the unborn child.
Toxic if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rin
EINECSNumber 203-804-1

RUO – Research Use Only