missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(Ethoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride), TCI America™
$46.20 - $46.20
Chemical Identifiers
| CAS | 380430-53-5 |
|---|---|
| Molecular Formula | C9H11BO4 |
| Molecular Weight (g/mol) | 193.99 |
| MDL Number | MFCD02179453 |
| InChI Key | QZKVVOXAEBCLPZ-UHFFFAOYSA-N |
| PubChem CID | 2773405 |
| IUPAC Name | [2-(ethoxycarbonyl)phenyl]boronic acid |
| SMILES | CCOC(=O)C1=CC=CC=C1B(O)O |
Chemical Identifiers
| 380430-53-5 | |
| 193.99 | |
| QZKVVOXAEBCLPZ-UHFFFAOYSA-N | |
| [2-(ethoxycarbonyl)phenyl]boronic acid |
| C9H11BO4 | |
| MFCD02179453 | |
| 2773405 | |
| CCOC(=O)C1=CC=CC=C1B(O)O |
Specifications
| 380430-53-5 | |
| C9H11BO4 | |
| 1 g | |
| QZKVVOXAEBCLPZ-UHFFFAOYSA-N | |
| [2-(ethoxycarbonyl)phenyl]boronic acid | |
| 2773405 | |
| Crystalline Powder |
| 138°C | |
| MFCD02179453 | |
| 2-ethoxycarbonyl phenyl boronic acid, 2-ethoxycarbonylbenzeneboronic acid, 2-ethoxycarbonyl phenylboronic acid, 2-ethoxycarbonylphenyl boronic acid, o-ethoxycarbonylphenylboronic acid, 2-carbethoxyphenylboronic acid, ethyl 2-dihydroxyboranyl benzoate, ethyl 2-boronobenzoate, 2-ethoxycarbonyl benzeneboronic acid | |
| CCOC(=O)C1=CC=CC=C1B(O)O | |
| 193.99 | |
| 193.99 | |
| 2-(Ethoxycarbonyl)phenylboronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No