missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzoic Acid 98.0+%, TCI America™

Catalog No. E004425G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E004425G 25 g
E0044500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E004425G Supplier TCI America Supplier No. E004425G

Chemical Identifiers

CAS 134-11-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00002438
InChI Key XDZMPRGFOOFSBL-UHFFFAOYSA-N
Synonym o-ethoxybenzoic acid, benzoic acid, 2-ethoxy, benzoic acid, o-ethoxy, o-ethylsalicylic acid, benzoic acid, ethoxy, 2'-carboxyphenetole, 2-ethoxy benzoic acid, 2-ethoxy-benzoic acid, pubchem20122, ortho-ethoxybenzoic acid
PubChem CID 67252
IUPAC Name 2-ethoxybenzoic acid
SMILES CCOC1=CC=CC=C1C(O)=O

Specifications

Color Yellow
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethoxybenzoic Acid