missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzoic acid, 98%

CAS: 134-11-2 | C9H10O3 | 166.18 g/mol

Supplier:  Thermo Scientific Chemicals A1475822

 View more versions of this product

Catalog No. AAA1475822

Only null left
Explore available promotions

Description

Description

2-Ethoxybenzoic acid is used in thermally induced solvent-free reactions of triphenylbismuth with a series of thiols and carboxylic acids. It serves as an active component of dental cement. Further, it prevents the entry of root-canal pathogens into periapical regions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxybenzoic acid is used in thermally induced solvent-free reactions of triphenylbismuth with a series of thiols and carboxylic acids. It serves as an active component of dental cement. Further, it prevents the entry of root-canal pathogens into periapical regions.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

2-Ethoxybenzoic acid
19°C to 20°C
174°C to 175°C (15 mmHg)
C9H10O3
MFCD00002438
2691039
Soluble in water.
CCOC1=CC=CC=C1C(O)=O
166.18
166.18
134-11-2
1.105
>110°C (230°F)
1.541
100 g
o-ethoxybenzoic acid, benzoic acid, 2-ethoxy, benzoic acid, o-ethoxy, o-ethylsalicylic acid, benzoic acid, ethoxy, 2'-carboxyphenetole, 2-ethoxy benzoic acid, 2-ethoxy-benzoic acid, pubchem20122, ortho-ethoxybenzoic acid
XDZMPRGFOOFSBL-UHFFFAOYSA-N
2-ethoxybenzoic acid
67252
98%

Chemical Identifiers

134-11-2
166.18
XDZMPRGFOOFSBL-UHFFFAOYSA-N
67252
CCOC1=CC=CC=C1C(O)=O
C9H10O3
MFCD00002438
o-ethoxybenzoic acid, benzoic acid, 2-ethoxy, benzoic acid, o-ethoxy, o-ethylsalicylic acid, benzoic acid, ethoxy, 2'-carboxyphenetole, 2-ethoxy benzoic acid, 2-ethoxy-benzoic acid, pubchem20122, ortho-ethoxybenzoic acid
2-ethoxybenzoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 205-130-3

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only