missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxybenzoic acid, 98%

Catalog No. p-7022131
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1475814 25 g
AAA1475822 100 g
AAA1475836 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1475814 Supplier Thermo Scientific Chemicals Supplier No. A1475814

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 134-11-2 | C9H10O3 | 166.18 g/mol

2-Ethoxybenzoic acid is used in thermally induced solvent-free reactions of triphenylbismuth with a series of thiols and carboxylic acids. It serves as an active component of dental cement. Further, it prevents the entry of root-canal pathogens into periapical regions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethoxybenzoic acid is used in thermally induced solvent-free reactions of triphenylbismuth with a series of thiols and carboxylic acids. It serves as an active component of dental cement. Further, it prevents the entry of root-canal pathogens into periapical regions.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 134-11-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.18
MDL Number MFCD00002438
InChI Key XDZMPRGFOOFSBL-UHFFFAOYSA-N
Synonym o-ethoxybenzoic acid, benzoic acid, 2-ethoxy, benzoic acid, o-ethoxy, o-ethylsalicylic acid, benzoic acid, ethoxy, 2'-carboxyphenetole, 2-ethoxy benzoic acid, 2-ethoxy-benzoic acid, pubchem20122, ortho-ethoxybenzoic acid
PubChem CID 67252
IUPAC Name 2-ethoxybenzoic acid
SMILES CCOC1=CC=CC=C1C(O)=O

Specifications

Melting Point 19°C to 20°C
Density 1.105
Boiling Point 174°C to 175°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.541
Quantity 25 g
Beilstein 2691039
Solubility Information Soluble in water.
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 2-Ethoxybenzoic acid
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-130-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only