missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Ethoxy-3-methylpyrazine 98.0+%, TCI America™
$130.48 - $397.05
Chemical Identifiers
| CAS | 32737-14-7 |
|---|---|
| Molecular Formula | C7H10N2O |
| Molecular Weight (g/mol) | 138.17 |
| MDL Number | MFCD00038025 |
| InChI Key | MMKWCKGYULOKET-UHFFFAOYSA-N |
| PubChem CID | 122941 |
| IUPAC Name | 2-ethoxy-3-methylpyrazine |
| SMILES | CCOC1=NC=CN=C1C |
Chemical Identifiers
| 32737-14-7 | |
| 138.17 | |
| MMKWCKGYULOKET-UHFFFAOYSA-N | |
| 2-ethoxy-3-methylpyrazine |
| C7H10N2O | |
| MFCD00038025 | |
| 122941 | |
| CCOC1=NC=CN=C1C |
Specifications
| 32737-14-7 | |
| 88°C | |
| MFCD00038025 | |
| MMKWCKGYULOKET-UHFFFAOYSA-N | |
| 2-ethoxy-3-methylpyrazine | |
| 122941 | |
| ≥98.0% (GC,T) | |
| 2-Ethoxy-3-methylpyrazine |
| Colorless | |
| C7H10N2O | |
| 5 g | |
| CCOC1=NC=CN=C1C | |
| 138.17 | |
| 138.17 | |
| Liquid |
Safety and Handling
TSCA : Yes