accessibility menu, dialog, popup

UserName

CAS RN 32737-14-7

CH3ONNH3C

CAS RN 32737-14-7

IUPAC Name: 2-ethoxy-3-methylpyrazine
Synonyms: pyrazine, 2-ethoxy-3-methyl-
Molecular Weight (g/mol): 138.17
Molecular Formula: C7H10N2O
InChi Key: MMKWCKGYULOKET-UHFFFAOYSA-N
SMILES: CCOC1=NC=CN=C1C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results