missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxy-3-methylpyrazine 98.0+%, TCI America™

Catalog No. M08095G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M08095G 5 g
M080925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M08095G Supplier TCI America Supplier No. M08095G
Only null left

Chemical Identifiers

CAS 32737-14-7
Molecular Formula C7H10N2O
Molecular Weight (g/mol) 138.17
MDL Number MFCD00038025
InChI Key MMKWCKGYULOKET-UHFFFAOYSA-N
PubChem CID 122941
IUPAC Name 2-ethoxy-3-methylpyrazine
SMILES CCOC1=NC=CN=C1C

Specifications

Color Colorless
Boiling Point 88°C
Quantity 5 g
Formula Weight 138.17
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Ethoxy-3-methylpyrazine